Ethyl-DL-leucate 95%

Lieferant: Apollo Scientific

Synonyme: DL-Leucic Acid Ethyl Ester, Ethyl DL-2-Hydroxy-4-methylvalerate, DL-2-Hydroxy-4-methylvaleric Acid Ethyl Ester

OR953661-25G OR953661-5G
APOSOR953661-25GEA 464 EUR
APOSOR953661-25G APOSOR953661-5G
Ethyl-DL-leucate 95%
Ethyl-DL-leucate
Formel: C₈H₁₆O₃
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 10348-47-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR