4,5-Dichlorchinazolin 95%

Lieferant: Apollo Scientific

OR953494-1G OR953494-250MG
APOSOR953494-1GEA 126 EUR
APOSOR953494-1G APOSOR953494-250MG
4,5-Dichlorchinazolin 95%
4,5-Dichloroquinazoline
Formel: C₈H₄Cl₂N₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09909637
CAS-Nummer: 2148-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR