N-Boc-4-(fmoc-amino)piperidine-4-carboxylic acid 97%

Lieferant: Apollo Scientific

Synonyme: 1-Boc-4-(Fmoc-amino)-piperidine-4-carboxylic acid, N-Boc-4-(Fmoc-amino)piperidine-4-carboxylic acid

OR953106-1G OR953106-500MG OR953106-5G
APOSOR953106-1GEA 256 EUR
APOSOR953106-1G APOSOR953106-500MG APOSOR953106-5G
N-Boc-4-(fmoc-amino)piperidine-4-carboxylic acid 97%
N-Boc-4-(fmoc-amino)piperidine-4-carboxylic acid
Formel: C₂₆H₃₀N₂O₆
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01318741
CAS-Nummer: 183673-66-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR