Methyl(3-methylthiophen-2-ylmethyl)amine hydrochloride 98%

Lieferant: Apollo Scientific

OR952540-10G OR952540-1G OR952540-5G
APOSOR952540-10GEA 502 EUR
APOSOR952540-10G APOSOR952540-1G APOSOR952540-5G
Methyl(3-methylthiophen-2-ylmethyl)amine hydrochloride 98%
Methyl(3-methylthiophen-2-ylmethyl)amine hydrochloride
Formel: C₇H₁₂ClNS
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07106801
CAS-Nummer: 860710-37-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR