(1-Aminomethylcyclohexyl)dimethylamine 98%

Lieferant: Apollo Scientific

OR951791-1G OR951791-5G
APOSOR951791-1GEA 248 EUR
APOSOR951791-1G APOSOR951791-5G
(1-Aminomethylcyclohexyl)dimethylamine 98%
(1-Aminomethylcyclohexyl)dimethylamine
Formel: C₉H₂₀N₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD05201342
CAS-Nummer: 41806-09-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR