3,4-Dihydro-1H-quinoxalin-2-one 95%
Lieferant: Apollo Scientific
Synonyme:
3,4-Dihydroquinoxalin-2(1H)-one
OR951424-1G
OR951424-5G
APOSOR951424-1GEA
55.1
EUR
APOSOR951424-1G
APOSOR951424-5G
3,4-Dihydro-1H-quinoxalin-2-one 95%
3,4-Dihydro-1H-quinoxalin-2-one
Formel:
C₈H₈N₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00204133 CAS-Nummer: 59564-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...