4-Methyl-1H-benzimidazole-2-carboxylic acid 95%
Lieferant: Apollo Scientific
OR951085-1G
OR951085-250MG
OR951085-5G
APOSOR951085-1GEA
132
EUR
APOSOR951085-1G
APOSOR951085-250MG
APOSOR951085-5G
4-Methyl-1H-benzimidazole-2-carboxylic acid 95%
4-Methyl-1H-benzimidazole-2-carboxylic acid
Formel:
C₉H₈N₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09751735 CAS-Nummer: 673487-32-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...