1,5-Naphthyridin-4-ol 95%

Lieferant: Apollo Scientific

OR951046-1G OR951046-250MG
APOSOR951046-1GEA 155 EUR
APOSOR951046-1G APOSOR951046-250MG
1,5-Naphthyridin-4-ol 95%
1,5-Naphthyridin-4-ol
Formel: C₈H₆N₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00234402
CAS-Nummer: 5423-54-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR