4-Methyl-2-thiazolecarbonitrile 95%
Lieferant: Apollo Scientific
OR948386-1G
OR948386-250MG
APOSOR948386-1GEA
538
EUR
APOSOR948386-1G
APOSOR948386-250MG
4-Methyl-2-thiazolecarbonitrile 95%
4-Methyl-2-thiazolecarbonitrile
Formel:
C₅H₄N₂S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08276856 CAS-Nummer: 100516-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...