Ethyl trans-β-methylcinnamate 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl trans-beta-methylcinnamate, Ethyl (2E)-3-phenyl-2-butenoate
OR945105-1G
OR945105-5G
APOSOR945105-1GEA
168
EUR
APOSOR945105-1G
APOSOR945105-5G
Ethyl trans-β-methylcinnamate 95%
Ethyl trans-β-methylcinnamate
Formel:
C₁₂H₁₄O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00053762 CAS-Nummer: 1504-72-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...