(±)-3-Methylcyclohexanon 95%
Lieferant: Apollo Scientific
OR939805-100G
OR939805-25G
APOSOR939805-100GEA
219
EUR
APOSOR939805-100G
APOSOR939805-25G
(±)-3-Methylcyclohexanon 95%
(±)-3-Methylcyclohexanon
Formel:
C₇H₁₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00001639 CAS-Nummer: 591-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...