4-Isobutylanilin 95%

Lieferant: Apollo Scientific

OR939250-1G OR939250-250MG
APOSOR939250-1GEA 192 EUR
APOSOR939250-1G APOSOR939250-250MG
4-Isobutylanilin 95%
4-Isobutylanilin
Formel: C₁₀H₁₅N
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00130035
CAS-Nummer: 30090-17-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR