Ethyl 2-Isopropylacetoacetate 90%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-Acetyl-3-methylbutyrate, 2-Acetyl-3-methylbutyric acid Ethyl ester, 2-Isopropylacetoacetic acid Ethyl ester

OR938251-1G OR938251-25G OR938251-5G
APOSOR938251-1GEA 102 EUR
APOSOR938251-1G APOSOR938251-25G APOSOR938251-5G
Ethyl 2-Isopropylacetoacetate 90%
Ethyl 2-Isopropylacetoacetate
Formel: C₉H₁₆O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00042884
CAS-Nummer: 1522-46-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR