2-Chlorophenylacetyl chloride 95%

Lieferant: Apollo Scientific

OR938171-1G OR938171-5G
APOSOR938171-1GEA 25.8 EUR
APOSOR938171-1G APOSOR938171-5G
2-Chlorophenylacetyl chloride 95%
2-Chlorophenylacetyl chloride
Formel: C₈H₆Cl₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00130223
CAS-Nummer: 51512-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR