2,3-Dichloropropionamide 95%
Lieferant: Apollo Scientific
OR938050-10G
OR938050-1G
OR938050-5G
APOSOR938050-10GEA
647
EUR
APOSOR938050-10G
APOSOR938050-1G
APOSOR938050-5G
2,3-Dichloropropionamide 95%
2,3-Dichloropropionamide
Formel:
C₃H₅Cl₂NO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00040311 CAS-Nummer: 19433-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...