2,2-dimethyl-3,4-dihydro-2H-1,4-benzoxazine 97%
Lieferant: Apollo Scientific
Synonyme:
2,2-Dimethyl-3,4-dihydro-2H-benzo[b][1,4]oxazine
OR937981-1G
OR937981-250MG
APOSOR937981-1GEA
231
EUR
APOSOR937981-1G
APOSOR937981-250MG
2,2-dimethyl-3,4-dihydro-2H-1,4-benzoxazine 97%
2,2-dimethyl-3,4-dihydro-2H-1,4-benzoxazine
Formel:
C₁₀H₁₃NO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11603419 CAS-Nummer: 866089-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...