1-(3-Hydroxypropyl)pyrrole 95%
Lieferant: Apollo Scientific
Synonyme:
3-(1H-Pyrrol-1-yl)propan-1-ol
OR937967-1G
OR937967-250MG
APOSOR937967-1GEA
128
EUR
APOSOR937967-1G
APOSOR937967-250MG
1-(3-Hydroxypropyl)pyrrole 95%
1-(3-Hydroxypropyl)pyrrole
Formel:
C₇H₁₁NO Molekulargewicht: 125.17 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00191453 CAS-Nummer: 50966-69-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...