N-Phenylhydroxylamine 95%

Lieferant: Apollo Scientific

OR933342-1G OR933342-250MG OR933342-5G
APOSOR933342-1GEA 248 EUR
APOSOR933342-1G APOSOR933342-250MG APOSOR933342-5G
N-Phenylhydroxylamine 95%
N-Phenylhydroxylamine
Formel: C₆H₇NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00045718
CAS-Nummer: 100-65-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR