4-(2-Thienyl)pyrimidine-2-thiol

Lieferant: Apollo Scientific

OR9331-1G OR9331-250MG OR9331-500MG OR9331-5G OR9331-10G OR9331-25G
APOSOR9331-1GEA 78.3 EUR
APOSOR9331-1G APOSOR9331-250MG APOSOR9331-500MG APOSOR9331-5G APOSOR9331-10G APOSOR9331-25G
4-(2-Thienyl)pyrimidine-2-thiol
4-(2-Thienyl)pyrimidine-2-thiol
Formel: C₈H₆N₂S₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00085173
CAS-Nummer: 175202-75-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR