Methoxymethyl-phenylsulfid 95%

Lieferant: Apollo Scientific

Synonyme: alpha-Methoxythioanisole, Methyl Phenylthiomethyl Ether

OR932563-10G OR932563-1G OR932563-5G
APOSOR932563-10GEA 290 EUR
APOSOR932563-10G APOSOR932563-1G APOSOR932563-5G
Methoxymethyl-phenylsulfid 95%
Methoxymethyl phenyl sulphide
Formel: C₈H₁₀OS
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00008496
CAS-Nummer: 13865-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR