2-(2-Methylphenyl)ethanol 97%

Lieferant: Apollo Scientific

OR931809-1G OR931809-5G
APOSOR931809-1GEA 21.5 EUR
APOSOR931809-1G APOSOR931809-5G
2-(2-Methylphenyl)ethanol 97%
2-(2-Methylphenyl)ethanol
Formel: C₉H₁₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00044241
CAS-Nummer: 19819-98-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR