5-Chloro-1-methylisatin 95%

Lieferant: Apollo Scientific

OR931368-1G OR931368-5G
APOSOR931368-1GEA 85.9 EUR
APOSOR931368-1G APOSOR931368-5G
5-Chloro-1-methylisatin 95%
Formel: C₉H₆ClNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02956479
CAS-Nummer: 60434-13-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR