1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine 95%

Lieferant: Apollo Scientific

OR930876-100G OR930876-25G
APOSOR930876-100GEA 228 EUR
APOSOR930876-100G APOSOR930876-25G
1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine 95%
1-(tert-Butoxycarbonyl)-3-(cyanomethylene)azetidine
Formel: C₁₀H₁₄N₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12761241
CAS-Nummer: 1153949-11-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR