N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(tert-Butoxycarbonylamino)-2-phenylethanol, N-Boc-DL-2-phenylglycinol, Boc-DL-Phenylglycinol, tert-Butyl (2-hydroxy-1-phenylethyl)carbamate

OR929831-1G OR929831-5G
APOSOR929831-1GEA 21.9 EUR
APOSOR929831-1G APOSOR929831-5G
N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol 97%
N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol
Formel: C₁₃H₁₉NO₃
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01883039
CAS-Nummer: 67341-01-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR