Fmoc-val-OSu 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Fmoc-L-valine N-succinimidyl ester, Fmoc-Val-OSu

OR928600-25G OR928600-100G
APOSOR928600-25GEA 34.8 EUR
APOSOR928600-25G APOSOR928600-100G
Fmoc-val-OSu 95%
Fmoc-val-OSu
Formel: C₂₄H₂₄N₂O₆
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00153370
CAS-Nummer: 130878-68-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR