C-(3,5-Dimethyl-1-phenyl-1H-pyrazol-4-yl)methylamine 95%
Lieferant: Apollo Scientific
Synonyme:
4-(Aminomethyl)-3,5-dimethyl-1-phenyl-1H-pyrazole hydrochloride
OR9270-1G
OR9270-250MG
APOSOR9270-1GEA
460
EUR
APOSOR9270-1G
APOSOR9270-250MG
C-(3,5-Dimethyl-1-phenyl-1H-pyrazol-4-yl)methylamine 95%
C-(3,5-Dimethyl-1-phenyl-1H-pyrazol-4-yl)methylamine
Formel:
C₁₂H₁₅N₃ Schmelzpunkt: 192…192.5 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02055795 CAS-Nummer: 879896-52-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...