4-Acetamidobenzoyl chloride 95%

Lieferant: Apollo Scientific

OR926403-1G OR926403-5G
APOSOR926403-1GEA 109 EUR
APOSOR926403-1G APOSOR926403-5G
4-Acetamidobenzoyl chloride 95%
4-Acetamidobenzoyl chloride
Formel: C₉H₈ClNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02094022
CAS-Nummer: 16331-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR