Dimethylphenylsilanol 95%
Lieferant: Apollo Scientific
OR926383-1G
OR926383-5G
APOSOR926383-1GEA
139
EUR
APOSOR926383-1G
APOSOR926383-5G
Dimethylphenylsilanol 95%
Dimethylphenylsilanol
Formel:
C₈H₁₂OSi Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08276879 CAS-Nummer: 5272-18-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...