(3-Benzyloxypropyl)triphenylphosphoniumbromid 97%

Lieferant: Apollo Scientific

Synonyme: [3-(Benzyloxy)propyl]triphenylphosphonium Bromide

OR926016-1G OR926016-5G
APOSOR926016-1GEA 72.5 EUR
APOSOR926016-1G APOSOR926016-5G
(3-Benzyloxypropyl)triphenylphosphoniumbromid 97%
(3-Benzyloxypropyl)triphenylphosphoniumbromid
Formel: C₂₈H₂₈BrOP
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00191781
CAS-Nummer: 54314-85-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR