2',4',5'-Trimethylacetophenon 95%

Lieferant: Apollo Scientific

Synonyme: 1-(2,4,5-Trimethylphenyl)ethan-1-one, 1-(2,4,5-Trimethylphenyl)ethanone

OR924731-25G OR924731-5G
APOSOR924731-25GEA 328 EUR
APOSOR924731-25G APOSOR924731-5G
2',4',5'-Trimethylacetophenon 95%
2',4',5'-Trimethylacetophenon
Formel: C₁₁H₁₄O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00017244
CAS-Nummer: 2040-07-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR