2-(Trimethylsilylethinyl)pyridin 95%

Lieferant: Apollo Scientific

Synonyme: (2-Pyridylethynyl)trimethylsilane, 2-[(Trimethylsilyl)ethynyl]pyridine, 2-((Trimethylsilyl)ethynyl)pyridine

OR923795-25G OR923795-5G
APOSOR923795-25GEA 289 EUR
APOSOR923795-25G APOSOR923795-5G
2-(Trimethylsilylethinyl)pyridin 95%
2-(Trimethylsilylethinyl)pyridin
Formel: C₁₀H₁₃NSi
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00066349
CAS-Nummer: 86521-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR