1-Chlor-2-methylpentan 98%
Lieferant: Apollo Scientific
OR923251-1G
OR923251-25G
OR923251-5G
APOSOR923251-1GEA
43.5
EUR
APOSOR923251-1G
APOSOR923251-25G
APOSOR923251-5G
1-Chlor-2-methylpentan 98%
1-Chloro-2-methylpentane
Formel:
C₆H₁₃Cl Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00053236 CAS-Nummer: 14753-05-0 UN-Nummer: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...