4-Methylanisol 95%
Lieferant: Apollo Scientific
Synonyme:
p-Methylanisole
OR922661-1KG
OR922661-500G
APOSOR922661-1KGEA
270
EUR
APOSOR922661-1KG
APOSOR922661-500G
4-Methylanisol 95%
4-Methylanisol
Formel:
C₈H₁₀O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00008413 CAS-Nummer: 104-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...