N,N-Diethyl-2-pyridinamin 95%
Lieferant: Apollo Scientific
Synonyme:
N-(2-Pyridyl)diethylamine
OR922656-1G
OR922656-25G
OR922656-5G
APOSOR922656-1GEA
154
EUR
APOSOR922656-1G
APOSOR922656-25G
APOSOR922656-5G
N,N-Diethyl-2-pyridinamin 95%
2-Diethylaminopyridine
Formel:
C₉H₁₄N₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00059777 CAS-Nummer: 36075-06-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...