Sodium hydroxymethanesulphinate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Sodium formaldehydesulphoxylate
OR922468-100G
OR922468-25G
APOSOR922468-100GEA
75.9
EUR
APOSOR922468-100G
APOSOR922468-25G
Sodium hydroxymethanesulphinate 95%
Sodium hydroxymethanesulphinate
Formel:
CH₃NaO₃S Schmelzpunkt: >51 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00040426 CAS-Nummer: 149-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...