5-Ethyl-2-furaldehyd 95%

Lieferant: Apollo Scientific

Synonyme: 5-Ethylfurfural

OR922057-1G OR922057-5G
APOSOR922057-1GEA 145 EUR
APOSOR922057-1G APOSOR922057-5G
5-Ethyl-2-furaldehyd 95%
5-Ethyl-2-furaldehyd
Formel: C₇H₈O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00030605
CAS-Nummer: 23074-10-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR