3-Methyl-5-phenylpyridin 95%

Lieferant: Apollo Scientific

OR922029-1G OR922029-5G
APOSOR922029-1GEA 116 EUR
APOSOR922029-1G APOSOR922029-5G
3-Methyl-5-phenylpyridin 95%
3-Methyl-5-phenylpyridin
Formel: C₁₂H₁₁N
Molekulargewicht: 169.23 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00191465
CAS-Nummer: 10477-94-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR