2-Ethylanisol 95%

Lieferant: Apollo Scientific

Synonyme: 1-Ethyl-2-methoxybenzene, 2-Ethylmethoxybenzene

OR921671-25G OR921671-5G
APOSOR921671-25GEA 264 EUR
APOSOR921671-25G APOSOR921671-5G
2-Ethylanisol 95%
2-Ethylanisol
Formel: C₉H₁₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00142909
CAS-Nummer: 14804-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR