Methyl-6-hydroxy-2-naphthoat 97%

Lieferant: Apollo Scientific
Achtung

OR921588-1G OR921588-25G OR921588-5G
APOSOR921588-1GEA 24.4 EUR
APOSOR921588-1G APOSOR921588-25G APOSOR921588-5G
Methyl-6-hydroxy-2-naphthoat 97%
Methyl-6-hydroxy-2-naphthoat
Formel: C₁₂H₁₀O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00100526
CAS-Nummer: 17295-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR