6-Methyl-5-nitroisoquinoline 95%

Lieferant: Apollo Scientific

OR921242-1G OR921242-250MG
APOSOR921242-1GEA 88.6 EUR
APOSOR921242-1G APOSOR921242-250MG
6-Methyl-5-nitroisoquinoline 95%
6-Methyl-5-nitroisoquinoline
Formel: C₁₀H₈N₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11848649
CAS-Nummer: 188121-31-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR