S-Methyl-2-furancarbothioate 98%

Lieferant: Apollo Scientific

OR921105-1G OR921105-25G OR921105-5G
APOSOR921105-1GEA 0 EUR
APOSOR921105-1G APOSOR921105-25G APOSOR921105-5G
S-Methyl-2-furancarbothioate 98%
S-Methyl-2-furancarbothioate
Formel: C₆H₆O₂S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00040266
CAS-Nummer: 13679-61-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR