Ethyl-5-chloropyrazine-2-carboxylate 95%
Lieferant: Apollo Scientific
OR920766-1G
OR920766-250MG
APOSOR920766-1GEA
88.6
EUR
APOSOR920766-1G
APOSOR920766-250MG
Ethyl-5-chloropyrazine-2-carboxylate 95%
Ethyl-5-chloropyrazine-2-carboxylate
Formel:
C₇H₇ClN₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD18070850 CAS-Nummer: 54013-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...