1,2-Epoxyheptan 95%

Lieferant: Apollo Scientific

OR919532-1G OR919532-5G
APOSOR919532-1GEA 219 EUR
APOSOR919532-1G APOSOR919532-5G
1,2-Epoxyheptan 95%
1,2-Epoxyheptane
Formel: C₇H₁₄O
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 5063-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR