Fmoc-D-Bpa-OH 97%
Lieferant: Apollo Scientific
Synonyme:
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-benzoylphenyl)propanoic acid, Fmoc-p-Bz-D-Phe-OH, N-Fmoc-4-benzoyl-D-phenylalanine
OR919316-1G
OR919316-500MG
OR919316-5G
APOSOR919316-1GEA
166
EUR
APOSOR919316-1G
APOSOR919316-500MG
APOSOR919316-5G
Fmoc-D-Bpa-OH 97%
Fmoc-D-Bpa-OH
Formel:
C₃₁H₂₅NO₅ Molekulargewicht: 491.543 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00237666 CAS-Nummer: 117666-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...