4'-Chloro-N-methylformanilide 98%
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-N-formyl-N-methylaniline
OR919092-1G
OR919092-25G
OR919092-5G
APOSOR919092-1GEA
168
EUR
APOSOR919092-1G
APOSOR919092-25G
APOSOR919092-5G
4'-Chloro-N-methylformanilide 98%
4'-Chloro-N-methylformanilide
Formel:
C₈H₈ClNO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02093455 CAS-Nummer: 26772-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...