2-Amino-3-(naphthalen-1-yl)propanoic acid 97%
Lieferant: Apollo Scientific
Synonyme:
H-DL-1-Nal-OH, 3-(1-Naphthyl)-DL-alanine
OR918989-1G
OR918989-25G
OR918989-5G
APOSOR918989-1GEA
106
EUR
APOSOR918989-1G
APOSOR918989-25G
APOSOR918989-5G
2-Amino-3-(naphthalen-1-yl)propanoic acid 97%
2-Amino-3-(naphthalen-1-yl)propanoic acid
Formel:
C₁₃H₁₃NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00067507 CAS-Nummer: 28095-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...