2-Bromobutyryl chloride 95%

Lieferant: Apollo Scientific

OR918969-25G OR918969-5G
APOSOR918969-25GEA 116 EUR
APOSOR918969-25G APOSOR918969-5G
2-Bromobutyryl chloride 95%
2-Bromobutyryl chloride
Formel: C₄H₆BrClO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00055269
CAS-Nummer: 22118-12-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR