N-tert-Butoxycarbonylimidazole 98%
Lieferant: Apollo Scientific
OR918828-100G
OR918828-25G
OR918828-500G
APOSOR918828-100GEA
43
EUR
APOSOR918828-100G
APOSOR918828-25G
APOSOR918828-500G
N-tert-Butoxycarbonylimidazole 98%
N-tert-Butoxycarbonylimidazole
Formel:
C₈H₁₂N₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00014497 CAS-Nummer: 49761-82-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...