N-Phenylmethansulfonamid 98%
Lieferant: Apollo Scientific
Synonyme:
N-(Methylsulphonyl)aniline
OR918539-100G
OR918539-25G
APOSOR918539-100GEA
284
EUR
APOSOR918539-100G
APOSOR918539-25G
N-Phenylmethansulfonamid 98%
N-Phenylmethansulfonamid
Formel:
C₇H₉NO₂S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00043782 CAS-Nummer: 1197-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...