Isopropyl glycolate 95%

Lieferant: Apollo Scientific

OR918109-1G OR918109-5G
APOSOR918109-1GEA 27.3 EUR
APOSOR918109-1G APOSOR918109-5G
Isopropyl glycolate 95%
Isopropyl glycolate
Formel: HOCH₂CO₂CH(CH₃)₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00046747
CAS-Nummer: 623-61-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR